3-Idrossichinolina ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₉H₇NO |
MDL Number:
MFCD00169018 CAS Number: 580-18-7 EINECS: 209-456-7 |
|
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 201.0 to 205.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...