3-Quinolinecarboxylic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₀H₇NO₂ Punto di fusione: 280°C |
MDL Number:
MFCD00006770 CAS Number: 6480-68-8 EINECS: 229-337-3 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...