Quincoridine ≥95.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₀H₁₇NO Flash Pt: 122°C |
CAS Number:
207129-36-0 |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Specific rotation [a]20/D | +190 to +200 deg (C=1, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...