7-Quinolinecarboxaldehyde ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₀H₇NO |
MDL Number:
MFCD01366121 CAS Number: 49573-30-0 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 85.0 to 88.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...