Quinacridone ≥93.0% (by total nitrogen basis)
Fornitore: TCI
Synonyms:
5,12-Dihydroquino[2,3-b]acridine-7,14-dione, Pigment Violet 19
Formula:
C₂₀H₁₂N₂O₂ Punto di fusione: 390°C |
MDL Number:
MFCD00059956 CAS Number: 1047-16-1 EINECS: 213-879-2 |
|
Risultati del test di specificazione
Appearance | Red to Dark red to Dark purple powder to crystal |
Purity (with Total Nitrogen) | min. 93.0 % |
Lambda max. | 596.0 to 600.0 nm (H2SO4) |
Absorbance (E1%1cm) | min. 300 (596.0 to 600.0 nm, H2SO4) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...