Stampa…

p-Quinone-4-chloroimide ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: N-Chloro-p-benzoquinone Monoimine, N-Chloro-p-quinone Monoimine

Q0019-1G Q0019-5G
TCIAQ0019-1GEA 78 EUR
TCIAQ0019-1G TCIAQ0019-5G
p-Quinone-4-chloroimide ≥95.0% (by titrimetric analysis)
p-Quinone-4-chloroimide
Formula: C₆H₄ClNO
Peso molecolare: 141,55 g/mol
MDL Number: MFCD00058961
CAS Number: 637-61-6
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5392153ac643e5216fdf92079595f2410e2bb058&vid=5392153ac643e5216fdf92079595f2410e2bb058&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (Iodometric Titration) min. 95.0 %
Melting point 82.0 to 86.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR