Stampa…

p-Quinquephenyl

Fornitore: TCI EUROPE

Q0018-1G Q0018-100MG
TCIAQ0018-1GEA 545 EUR
TCIAQ0018-1G TCIAQ0018-100MG
p-Quinquephenyl
p-Quinquephenyl
Formula: C₃₀H₂₂
MDL Number: MFCD00059012
CAS Number: 3073-05-0

Visualizza i codici

Risultati del test di specificazione

Appearance White powder to crystal
TOF-MS to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR