Stampa…

8-Quinolinol sulphate monohydrate ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 8-Hydroxyquinoline sulphate monohydrate, Chinosol monohydrate, 8-Quinolinone sulphate monohydrate, Oxine sulphate monohydrate

Q0017-25G Q0017-500G
TCIAQ0017-25GEA 23 EUR
TCIAQ0017-25G TCIAQ0017-500G
8-Quinolinol sulphate monohydrate ≥98.0% (by titrimetric analysis)
8-Quinolinol sulphate monohydrate
Formula: C₁₈H₁₄N₂O₂·H₂SO₄·H₂O
Peso molecolare: 406,41 g/mol
Punto di fusione: 178°C
MDL Number: MFCD00013095
CAS Number: 134-31-6
EINECS: 205-137-1
Merck Index: 14,04843

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water 4.0 to 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR