Polyethylene glycol diacrylate (PEGDA) n=∼14 stabilizzato
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
PEG hydrogel, Polyethylene glycol diacrylate, Poly(ethylene glycol) diacrylate, PEG diacrylate, PEGDA, PEG bisacrylate
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
P2757-25G
P2757-500G
TCIAP2757-25GEA
19
EUR
TCIAP2757-25G
TCIAP2757-500G
Polyethylene glycol diacrylate (PEGDA) n=∼14 stabilizzato
Polyethylene glycol diacrylate (PEGDA)
Formula:
CH₂=COC(OCH₂CH₂)nOCOC=CH₂ Flash Pt: 230°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00081876 CAS Number: 26570-48-9 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
The average value of n | 13.0 to 15.0 |
Specific gravity (20/20) | 1.1200 to 1.1300 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...