Stampa…

Polyethylene glycol diacrylate (PEGDA) n=∼4 stabilizzato

Fornitore: TCI EUROPE
Pericolo

Synonyms: PEG hydrogel, Polyethylene glycol diacrylate, Poly(ethylene glycol) diacrylate, PEG diacrylate, PEGDA, PEG bisacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

P2756-25G P2756-500G
TCIAP2756-25GEA 19 EUR
TCIAP2756-25G TCIAP2756-500G
Polyethylene glycol diacrylate (PEGDA) n=∼4 stabilizzato
Polyethylene glycol diacrylate (PEGDA)
Formula: CH₂=COC(OCH₂CH₂)nOCOC=CH₂
Flash Pt: 104°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00081876
CAS Number: 26570-48-9

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
The average value of n 3.5 to 4.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR