Stampa…

(+/-)-Phenyl(pyridin-2-yl)methanol ≥98.0% (by GC)

Fornitore: TCI

P2755-1G P2755-5G
TCIAP2755-1GEA 75 EUR
TCIAP2755-1G TCIAP2755-5G
(+/-)-Phenyl(pyridin-2-yl)methanol ≥98.0% (by GC)
(+/-)-Phenyl(pyridin-2-yl)methanol
Formula: C₁₂H₁₁NO
MDL Number: MFCD00186514
CAS Number: 14159-57-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR