Sphynolactone-7 ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
SPL 7, 4-Methyl-5-oxo-2,5-dihydrofuran-2-yl 4-[(4-Butoxyphenyl)sulphonyl]piperazine-1-carboxylate
P2745-25MG
P2745-5MG
TCIAP2745-25MGEA
154
EUR
TCIAP2745-25MG
TCIAP2745-5MG
Sphynolactone-7 ≥98.0% (by HPLC)
Sphynolactone-7
Formula:
C₂₀H₂₆N₂O₇S Peso molecolare: 438,50 g/mol |
CAS Number:
2305752-57-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Melting point | 193.0 to 197.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...