Stampa…

4'-Pentyl-4-biphenylboronic acid ≥98.0% (by HPLC, titration analysis) (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(4-Pentylphenyl)benzeneboronic acid, (4'-Pentyl-[1,1'-biphenyl]-4-yl)boronic acid

P2717-1G P2717-5G
TCIAP2717-1GEA 60 EUR
TCIAP2717-1G TCIAP2717-5G
4'-Pentyl-4-biphenylboronic acid ≥98.0% (by HPLC, titration analysis) (contains varying amounts of Anhydride)
4'-Pentyl-4-biphenylboronic acid
Formula: C₁₇H₂₁BO₂
Peso molecolare: 268,16 g/mol
MDL Number: MFCD07644474
CAS Number: 121554-18-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) gt. 98.0 to 106.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR