Stampa…

Pirodavir ≥95.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: Ethyl 4-[2-[1-(6-Methyl-3-pyridazinyl)-4-piperidinyl]ethoxy]benzoate, Ethyl 4-(2-(1-(6-methylpyridazin-3-yl)piperidin-4-yl)ethoxy)benzoate, 4-[2-[1-(6-Methyl-3-pyridazinyl)-4-piperidinyl]ethoxy]benzoic acid Ethyl ester

P2665-10MG P2665-50MG
TCIAP2665-10MGEA 126 EUR
TCIAP2665-10MG TCIAP2665-50MG
Pirodavir ≥95.0% (by HPLC)
Pirodavir
Formula: C₂₁H₂₇N₃O₃
Peso molecolare: 369,47 g/mol
Punto di fusione: 125°C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00866965
CAS Number: 124436-59-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 10.50 to 12.30 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR