Stampa…

4-Phenoxybenzylamine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (4-Phenoxyphenyl)methanamine

P2664-25G P2664-5G
TCIAP2664-25GEA 253 EUR
TCIAP2664-25G TCIAP2664-5G
4-Phenoxybenzylamine ≥98.0% (by GC)
4-Phenoxybenzylamine
Formula: C₁₃H₁₃NO
Peso molecolare: 199,25 g/mol
MDL Number: MFCD01310836
CAS Number: 107622-80-0

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR