1-Phenyl-3-(p-tolyl)-1H-pyrazole-4-carboxaldehyde ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
1-Phenyl-3-(p-tolyl)-1H-pyrazole-4-carbaldehyde
P2657-200MG
P2657-1G
TCIAP2657-200MGEA
23
EUR
TCIAP2657-200MG
TCIAP2657-1G
1-Phenyl-3-(p-tolyl)-1H-pyrazole-4-carboxaldehyde ≥98.0% (by GC)
1-Phenyl-3-(p-tolyl)-1H-pyrazole-4-carboxaldehyde
Formula:
C₁₇H₁₄N₂O Peso molecolare: 262,31 g/mol |
MDL Number:
MFCD00744772 CAS Number: 36640-52-5 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 117.0 to 121.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...