Stampa…

2,2'-(1,4-Phenylene)bis(4H-3,1-benzoxazin-4-one) ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,2'-(1,4-Phenylene)bis(4H-benzo[d][1,3]oxazin-4-one)

P2656-25G P2656-5G
TCIAP2656-25GEA 155 EUR
TCIAP2656-25G TCIAP2656-5G
2,2'-(1,4-Phenylene)bis(4H-3,1-benzoxazin-4-one) ≥98.0% (by HPLC, total nitrogen)
2,2'-(1,4-Phenylene)bis(4H-3,1-benzoxazin-4-one)
Formula: C₂₂H₁₂N₂O₄
Peso molecolare: 368,35 g/mol
MDL Number: MFCD00356293
CAS Number: 18600-59-4
EINECS: 418-280-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR