Stampa…

Potassium(2-naphthalene)trifluoroborate ≥98.0% (by HPLC)

Fornitore: TCI
Attenzione

P2655-25G P2655-5G
TCIAP2655-25GEA 302 EUR
TCIAP2655-25G TCIAP2655-5G
Potassium(2-naphthalene)trifluoroborate ≥98.0% (by HPLC)
Potassium(2-naphthalene)trifluoroborate
Formula: C₁₀H₇BF₃K
MDL Number: MFCD06659941
CAS Number: 668984-08-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR