Stampa…

Propyl Levulinate ≥98.0% (by GC)

Fornitore: TCI

P2631-5G P2631-25G
TCIAP2631-5GEA 49 EUR
TCIAP2631-5G TCIAP2631-25G
Propyl Levulinate ≥98.0% (by GC)
Propyl Levulinate
Formula: C₈H₁₄O₃
Punto di ebollizione: 221°C
MDL Number: MFCD16041935
CAS Number: 645-67-0

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Acid Value max. 2.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR