Psoralidin ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
3,9-Dihydroxy-2-(3-methylbut-2-en-1-yl)-6H-benzofuro[3,2-c]chromen-6-one
P2605-100MG
P2605-25MG
TCIAP2605-100MGEA
277
EUR
TCIAP2605-100MG
TCIAP2605-25MG
Psoralidin ≥98.0% (by HPLC)
Psoralidin
Formula:
C₂₀H₁₆O₅ Peso molecolare: 336,3 g/mol Punto di fusione: 315°C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD10566617 CAS Number: 18642-23-4 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...