Pyridine-2-carboxamidoxime ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Pyridylamidoxime, N'-Hydroxypicolinimidamide, 2-Pyridinecarboxamide Oxime, N-Hydroxypicolinimidamide
P2590-25G
P2590-5G
TCIAP2590-25GEA
119
EUR
TCIAP2590-25G
TCIAP2590-5G
Pyridine-2-carboxamidoxime ≥98.0% (by HPLC, titration analysis)
Pyridine-2-carboxamidoxime
Formula:
C₆H₇N₃O Peso molecolare: 137,14 g/mol |
MDL Number:
MFCD00085159 CAS Number: 1772-01-6 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 116.0 to 120.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...