Stampa…

Parecoxib ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: N-[[4-(5-Methyl-3-phenylisoxazol-4-yl)phenyl]sulphonyl]propionamide

P2579-250MG P2579-50MG
TCIAP2579-250MGEA 334 EUR
TCIAP2579-250MG TCIAP2579-50MG
Parecoxib ≥97.0% (by HPLC, titration analysis)
Parecoxib

This product is unavailable in the U.S.

Formula: C₁₉H₁₈N₂O₄S
Peso molecolare: 370,42 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD08141856
CAS Number: 198470-84-7
Merck Index: 14,07036

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 153.0 to 157.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR