Stampa…

PD 158780 ≥97.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: N4-(3-Bromophenyl)-N6-methylpyrido[3,4-d]pyrimidine-4,6-diamine

P2567-10MG P2567-50MG
TCIAP2567-10MGEA 117 EUR
TCIAP2567-10MG TCIAP2567-50MG
PD 158780 ≥97.0% (by HPLC)
PD 158780
Formula: C₁₄H₁₂BrN₅
Peso molecolare: 330,19 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00942160
CAS Number: 171179-06-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Nitrogen) 20.00 to 22.00 %
Melting point 174.0 to 178.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR