Stampa…

5-Pyrazolone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,4-Dihydro-3H-pyrazol-3-one, 1H-Pyrazol-5(4H)-one

P2563-200MG P2563-1G
TCIAP2563-200MGEA 21 EUR
TCIAP2563-200MG TCIAP2563-1G
5-Pyrazolone ≥98.0% (by GC)
5-Pyrazolone
Formula: C₃H₄N₂O
Peso molecolare: 84,08 g/mol
MDL Number: MFCD00127223
CAS Number: 137-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef3b55b374ae87813e5df4d48a16e4c2c1b7ae8d&vid=ef3b55b374ae87813e5df4d48a16e4c2c1b7ae8d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 160.0 to 164.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR