Pemirolast potassium ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₀H₇KN₆O Punto di fusione: 317°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD01690051 CAS Number: 100299-08-9 Merck Index: 14,07078 |
|
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...