Stampa…

2-Phenoxyphenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Phenoxybenzeneboronic acid

P2539-1G P2539-5G
TCIAP2539-1GEA 28 EUR
TCIAP2539-1G TCIAP2539-5G
2-Phenoxyphenylboronic acid (contains varying amounts of Anhydride)
2-Phenoxyphenylboronic acid
Formula: C₁₂H₁₁BO₃
Peso molecolare: 214,03 g/mol
Punto di fusione: 114°C
MDL Number: MFCD01001592
CAS Number: 108238-09-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR