Stampa…

Pneumocandin B0 ≥96.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: Pneumocandin B0, (10R,12S)-N-((2R,6S,9S,11R,12R,14aS,15S,20S,23S,25aS)-20-((R)-3-Amino-1-hydroxy-3-oxopropyl)-23-((1S,2S)-1,2-dihydroxy-2-(4-hydroxyphenyl)ethyl)-2,11,12,15-tetrahydroxy-6-((R)-1-hydroxyethyl)-5,8,14,19,22,25-hexaoxotetracosahydro-1H-dipyrrolo[2,1-c:2',1'-l][1,4,7,10,13,16]hexaazacyclohenicosin-9-yl)-10,12-dimethyltetradecanamide

P2534-10MG P2534-50MG
TCIAP2534-10MGEA 150 EUR
TCIAP2534-10MG TCIAP2534-50MG
Pneumocandin B0 ≥96.0% (by HPLC)
Pneumocandin B0
Formula: C₅₀H₈₀N₈O₁₇
Peso molecolare: 1065,23 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD28168025
CAS Number: 135575-42-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Elemental analysis (Nitrogen) 9.50 to 11.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR