Stampa…

Potassium (Phthalimidomethyl)trifluoroborate ≥98.0% (by total nitrogen basis)

Fornitore: TCI EUROPE
Attenzione

P2514-1G P2514-5G
TCIAP2514-1GEA 119 EUR
TCIAP2514-1G TCIAP2514-5G
Potassium (Phthalimidomethyl)trifluoroborate ≥98.0% (by total nitrogen basis)
Potassium (Phthalimidomethyl)trifluoroborate
Formula: C₉H₆BF₃KNO₂
Peso molecolare: 267,06 g/mol
Punto di fusione: 276°C
MDL Number: MFCD19000584
CAS Number: 1001671-72-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR