Stampa…

Potassium (Acetoxymethyl)trifluoroborate ≥96.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Potassium (acetoxymethyl)trifluoroborate

P2494-1G P2494-5G
TCIAP2494-1GEA 253 EUR
TCIAP2494-1G TCIAP2494-5G
Potassium (Acetoxymethyl)trifluoroborate ≥96.0% (by titrimetric analysis)
Potassium (Acetoxymethyl)trifluoroborate
Formula: C₃H₅BF₃KO₂
Peso molecolare: 179,97 g/mol
MDL Number: MFCD28023894
CAS Number: 910251-35-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e00ab42d01695727d3ddda25e9385bd43119595&vid=5e00ab42d01695727d3ddda25e9385bd43119595&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR