N-[2-(2-Pyridyldithio)ethyl]biotinamide ≥97.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
Biotin-[2-(pyridin-2-ylthio)ethylamide]
P2471-10MG
P2471-50MG
TCIAP2471-10MGEA
56
EUR
TCIAP2471-10MG
TCIAP2471-50MG
N-[2-(2-Pyridyldithio)ethyl]biotinamide ≥97.0% (by HPLC)
N-[2-(2-Pyridyldithio)ethyl]biotinamide
Formula:
C₁₇H₂₄N₄O₂S₃ Peso molecolare: 412,59 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD01320375 CAS Number: 112247-65-1 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Elemental analysis (Nitrogen) | 12.90 to 14.20 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...