Stampa…

Potassium trifluoro(prop-1-en-2-yl)borate ≥98.0% (W)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Potassium Isopropenyltrifluoroborate

P2456-1G P2456-5G
TCIAP2456-1GEA 28 EUR
TCIAP2456-1G TCIAP2456-5G
Potassium trifluoro(prop-1-en-2-yl)borate ≥98.0% (W)
Potassium trifluoro(prop-1-en-2-yl)borate
Formula: C3H5BF3K
Peso molecolare: 147,9763 g/mol
MDL Number: MFCD11052654
CAS Number: 395083-14-4
UN: 3261
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Gravimetric) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR