Stampa…

4-Pyrid-2-ylbenzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(2-Pyridyl)benzoic acid, 4-(Pyridin-2-yl)benzoic acid

P2455-1G P2455-5G
TCIAP2455-1GEA 93 EUR
TCIAP2455-1G TCIAP2455-5G
4-Pyrid-2-ylbenzoic acid ≥98.0% (by GC, titration analysis)
4-Pyrid-2-ylbenzoic acid
Formula: C₁₂H₉NO₂
Peso molecolare: 199,2100 g/mol
MDL Number: MFCD01830093
CAS Number: 4385-62-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 236.0 to 240.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR