2-Phenylnicotinic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Phenylnicotinic acid, 2-Phenylpyridine-3-carboxylic acid
P2452-200MG
P2452-1G
TCIAP2452-200MGEA
37
EUR
TCIAP2452-200MG
TCIAP2452-1G
2-Phenylnicotinic acid ≥98.0% (by GC, titration analysis)
2-Phenylnicotinic acid
Formula:
C₁₂H₉NO₂ Peso molecolare: 199,21 g/mol |
MDL Number:
MFCD03069417 CAS Number: 33421-39-5 EINECS: 251-510-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 167.0 to 171.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...