Stampa…

trans-4-(4-n-Propylcyclohexyl)benzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(trans-4-Propylcyclohexyl)benzoic acid, 4-(trans-4-n-Propylcyclohexyl)benzoic acid

P2438-5G P2438-25G
TCIAP2438-5GEA 50 EUR
TCIAP2438-5G TCIAP2438-25G
trans-4-(4-n-Propylcyclohexyl)benzoic acid ≥98.0% (by GC, titration analysis)
trans-4-(4-n-Propylcyclohexyl)benzoic acid
Formula: C₁₆H₂₂O₂
Peso molecolare: 246,35 g/mol
Punto di fusione: 206°C
MDL Number: MFCD06658177
CAS Number: 65355-29-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da2645266a40d9cf347b0e65bf24d354ffa3e0c6&vid=da2645266a40d9cf347b0e65bf24d354ffa3e0c6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Mesomorphic range 214.0 to 262.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR