Stampa…

4-(Pyrrolidin-1-yl)benzonitrile ≥96.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 4-(1-Pyrrolidinyl)benzonitrile, 1-(4-Cyanophenyl)pyrrolidine

P2428-1G P2428-5G
TCIAP2428-1GEA 52 EUR
TCIAP2428-1G TCIAP2428-5G
4-(Pyrrolidin-1-yl)benzonitrile ≥96.0% (by GC, titration analysis)
4-(Pyrrolidin-1-yl)benzonitrile
Formula: C₁₁H₁₂N₂
Peso molecolare: 172,23 g/mol
MDL Number: MFCD07368512
CAS Number: 10282-30-1
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7574a907fe053e64e4a05c200fa9cf6fa4521e7b&vid=7574a907fe053e64e4a05c200fa9cf6fa4521e7b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 81.0 to 85.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR