Stampa…

(2E,4E)-5-Phenyl-2,4-pentadienoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

P2422-5G P2422-25G
TCIAP2422-5GEA 225 EUR
TCIAP2422-5G TCIAP2422-25G
(2E,4E)-5-Phenyl-2,4-pentadienoic acid ≥98.0% (by GC, titration analysis)
(2E,4E)-5-Phenyl-2,4-pentadienoic acid
Formula: C₁₁H₁₀O₂
MDL Number: MFCD00014018
CAS Number: 28010-12-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=123c0a86773552829bd87caa74274a1938fc68aa&vid=123c0a86773552829bd87caa74274a1938fc68aa&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 166.0 to 170.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR