Stampa…

Pyridine-2,4-dicarboxylic acid ≥98.0% (by GC)

Fornitore: TCI
Attenzione

P2416-5G P2416-25G
TCIAP2416-5GEA 36 EUR
TCIAP2416-5G TCIAP2416-25G
Pyridine-2,4-dicarboxylic acid ≥98.0% (by GC)
Pyridine-2,4-dicarboxylic acid
Formula: C₇H₅NO₄
Punto di fusione: 248°C
MDL Number: MFCD00006296
CAS Number: 499-80-9
EINECS: 207-892-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=057c3af8f8d6a7745c6794a7e713055a3abf7860&vid=203ad19738a0eca88d7e3d23bcca489043a6f205&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR