Phthaloyl Amlodipine ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI
Synonyms:
3-Ethyl 5-Methyl 4-(2-Chlorophenyl)-2-[[2-(1,3-dioxoisoindolin-2-yl)ethoxy]methyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, 4-(2-Chlorophenyl)-2-[[2-(1,3-dihydro-1,3-dioxo-2H-isoindol-2-yl)ethoxy]methyl]-1,4-dihydro-6-methyl-3,5-pyridinedicarboxylic acid 3-Ethyl 5-Methyl ester, 3-Ethyl 5-methyl 4-(2-chlorophenyl)-2-((2-(1,3-dioxoisoindolin-2-yl)ethoxy)methyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, Amlodipine besilate impurity A
Formula:
C₂₈H₂₇ClN₂O₇ Peso molecolare: 538.98 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD07782096 CAS Number: 88150-62-3 EINECS: 413-410-3 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 150.0 to 154.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...