Stampa…

trans,trans-4'-Propyl-4-(p-tolyl)bicyclohexyl ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: trans,trans-4'-Propyl-4-(4-methylphenyl)bicyclohexyl, 1-Methyl-4-[(trans,trans)-4'-propyl[1,1'-bicyclohexyl]-4-yl]benzene

P2404-1G P2404-5G
TCIAP2404-1GEA 22 EUR
TCIAP2404-1G TCIAP2404-5G
trans,trans-4'-Propyl-4-(p-tolyl)bicyclohexyl ≥98.0% (by GC)
trans,trans-4'-Propyl-4-(p-tolyl)bicyclohexyl
Formula: C₂₂H₃₄
Peso molecolare: 298,51 g/mol
MDL Number: MFCD13182311
CAS Number: 84656-75-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d70cc82029a9db2ef8d729e0544d3cf5a877858&vid=7d70cc82029a9db2ef8d729e0544d3cf5a877858&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 107 to 182 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR