Stampa…

2-Phenylethylamine hydrobromide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Phenylethylammonium bromide, PEABr, Phenethylamine hydrobromide

P2388-1G P2388-5G
TCIAP2388-1GEA 39 EUR
TCIAP2388-1G TCIAP2388-5G
2-Phenylethylamine hydrobromide ≥98.0% (by HPLC, titration analysis)
2-Phenylethylamine hydrobromide
Formula: C₈H₁₁N·HBr
Peso molecolare: 202,10 g/mol
Punto di fusione: 259°C
CAS Number: 53916-94-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa41c5e425de22c361dd5e50812f00957000c1b9&vid=fa41c5e425de22c361dd5e50812f00957000c1b9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR