Stampa…

[5-Phenyl-3-isoxazolyl]methanol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: (5-Phenylisoxazol-3-yl)methanol, 3-Hydroxymethyl-5-phenylisoxazole

P2386-1G P2386-5G
TCIAP2386-1GEA 134 EUR
TCIAP2386-1G TCIAP2386-5G
[5-Phenyl-3-isoxazolyl]methanol ≥98.0% (by GC)
[5-Phenyl-3-isoxazolyl]methanol
Formula: C₁₀H₉NO₂
Peso molecolare: 175,1840 g/mol
MDL Number: MFCD01444104
CAS Number: 1619-37-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b09e605d4d3989a316b6fe0f1383795e09e49c0&vid=6b09e605d4d3989a316b6fe0f1383795e09e49c0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 100.0 to 104.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR