2,2-Dimethyl-N-pyridin-2-ylpropionamide ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₀H₁₄N₂O |
MDL Number:
MFCD00460277 CAS Number: 86847-59-8 |
|
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 71.0 to 75.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...