Stampa…

2-(Piperidin-4-yl)-1H-benzo[d]imidazole ≥98.0% (by GC)

Fornitore: TCI

Synonyms: 2-(4-Piperidinyl)benzimidazole

P2382-1G P2382-5G
TCIAP2382-1GEA 68 EUR
TCIAP2382-1G TCIAP2382-5G
2-(Piperidin-4-yl)-1H-benzo[d]imidazole ≥98.0% (by GC)
2-(Piperidin-4-yl)-1H-benzo[d]imidazole
Formula: C₁₂H₁₅N₃
MDL Number: MFCD01475843
CAS Number: 38385-95-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50731cfde4f3b46181c5f34e8040f8f1a0113445&vid=50731cfde4f3b46181c5f34e8040f8f1a0113445&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR