Stampa…

trans,trans-4-[(E)-1-Propenyl]-4'-propylbicyclohexyl ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: (trans,trans)-4-(Prop-1-en-1-yl)-4'-propyl-1,1'-bi(cyclohexane)

P2371-1G P2371-5G
TCIAP2371-1GEA 30 EUR
TCIAP2371-1G TCIAP2371-5G
trans,trans-4-[(E)-1-Propenyl]-4'-propylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-[(E)-1-Propenyl]-4'-propylbicyclohexyl
Formula: C₁₈H₃₂
Peso molecolare: 248,45 g/mol
MDL Number: MFCD12761279
CAS Number: 279246-65-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49ec6ac59d4f195965e30b62c408ae5374846a5e&vid=49ec6ac59d4f195965e30b62c408ae5374846a5e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR