Stampa…

Pyrilamine maleate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: Mepyramine maleate, Anisopyradamine, Minihist, Paraminyl maleate, N1-(4-Methoxybenzyl)-N2,N2-dimethyl-N1-(pyridin-2-yl)ethane-1,2-diamine maleate, Diaminide maleate

P2369-5G P2369-25G
TCIAP2369-5GEA 22 EUR
TCIAP2369-5G TCIAP2369-25G
Pyrilamine maleate ≥98.0% (by HPLC, titration analysis)
Pyrilamine maleate
Formula: C₂₁H₂₇N₃O₅
Peso molecolare: 401.463 g/mol
MDL Number: MFCD00069333
CAS Number: 59-33-6
EINECS: 200-422-7
Merck Index: 14,07984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5d4882157f4560106c9597a9ffbb1ac636e746a&vid=b5d4882157f4560106c9597a9ffbb1ac636e746a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 % (total of Ion-Pair)
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 100.0 to 104.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR