Piperlongumine ≥97.0% (by HPLC)
Fornitore: TCI
Formula:
C₁₇H₁₉NO₅ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00075706 CAS Number: 20069-09-4 |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Elemental analysis (Nitrogen) | 4.10 to 4.70 % |
Melting point | 123.0 to 127.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...