Stampa…

potassium trimethylacetate ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 2,2-Dimethylpropionic acid potassium salt, Pivalic acid potassium salt, Potassium trimethylacetate, Potassium Pivalate, Trimethylacetic acid potassium salt, Potassium 2,2-Dimethylpropionate

P2354-5G P2354-25G
TCIAP2354-5GEA 84 EUR
TCIAP2354-5G TCIAP2354-25G
potassium trimethylacetate ≥98.0% (by titrimetric analysis)
potassium trimethylacetate
Formula: C₅H₉KO₂
Peso molecolare: 140,223 g/mol
MDL Number: MFCD00671345
CAS Number: 19455-23-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00881e2b19eb0f8a5d062d70c3f8d06b02c44d81&vid=00881e2b19eb0f8a5d062d70c3f8d06b02c44d81&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR