Stampa…

9-Phenyl-9H-carbazole-3-carboxaldehyde ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 9-Phenyl-9H-carbazole-3-carbaldehyde, 3-Formyl-9-phenyl-9H-carbazole

P2351-1G P2351-5G
TCIAP2351-1GEA 127 EUR
TCIAP2351-1G TCIAP2351-5G
9-Phenyl-9H-carbazole-3-carboxaldehyde ≥98.0% (by GC)
9-Phenyl-9H-carbazole-3-carboxaldehyde
Formula: C₁₉H₁₃NO
Peso molecolare: 271,32 g/mol
MDL Number: MFCD19441087
CAS Number: 87220-68-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=912af51da12464fcf8698d09abbe862b36d1286f&vid=912af51da12464fcf8698d09abbe862b36d1286f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR