Stampa…

Pizotifen ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-[9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene]-1-methylpiperidine, Sandomigran, 4-(9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-ylidene)-1-methylpiperidine

P2344-50MG P2344-200MG
TCIAP2344-50MGEA 39 EUR
TCIAP2344-50MG TCIAP2344-200MG
Pizotifen ≥98.0% (by HPLC)
Pizotifen
Formula: C₁₉H₂₁NS
Peso molecolare: 295,44 g/mol
MDL Number: MFCD00864192
CAS Number: 15574-96-6
EINECS: 239-632-9
Merck Index: 14,07515
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4121ca3be4611a3ca57b832ec243e42505c49d3b&vid=4121ca3be4611a3ca57b832ec243e42505c49d3b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR