Stampa…

1-Cyanopiperidine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1-Cyanopiperidine, Pentamethylenecyanamide, Piperidine-1-carbonitrile

P2343-1G P2343-5G
TCIAP2343-1GEA 45 EUR
TCIAP2343-1G TCIAP2343-5G
1-Cyanopiperidine ≥98.0% (by GC)
1-Cyanopiperidine
Formula: C₆H₁₀N₂
Punto di ebollizione: 103°C
Flash Pt: 97°C
MDL Number: MFCD00006477
CAS Number: 1530-87-6
EINECS: 216-233-8
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=902dbcfe3d7ed22937608d04ed06345f34bd7b67&vid=012bea0059ab2e611756bd2555db9e18d08be9b5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR